Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RELT/TNFRSF19L Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1385G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1385G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1385G100 Supplier R&D Systems Supplier No. FAB1385G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RELT/TNFRSF19L Monoclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen RELT/TNFRSF19L
Applications Flow Cytometry
Classification Monoclonal
Clone 238104
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q969Z4
Gene Alias Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19L, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like
Gene Symbols RELT
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human RELT/TNFRSF19L Met1-Ala160 (Arg127Gly, Arg129Gly) Accession # Q969Z4
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84957
Test Specificity Detects human RELT/TNFRSF19L in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, recombinant mouse (rm) 4-1BB, rhBAFF R, rmBAFF R, rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rmDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFas, rrFas, rhGITR, rmGITR, rhHVEM, rhLT beta R, rmLT beta R, rhNGF R, rmNGF R, rhOPG, rmOPG, rmOX40, rhRANK, rmRANK, rhTNF RI, rmTNF RI, rhTNF RII, rmTNF RII, rhTRAIL R1, rhTRAIL R2, rhTRAIL R3, rhTRAIL R4, rhTROY, or rmTROY is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.