Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Resistin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1359
This item is not returnable.
View return policy
Description
Resistin Polyclonal specifically detects Resistin in Human samples. It is validated for Western Blot.Specifications
Resistin | |
Polyclonal | |
Unconjugated | |
Q9HD89 | |
RETN | |
E. coli-derived recombinant human Resistin Ser17-Pro108 Accession # Q9HD89 | |
100 μg | |
Primary | |
Detects human Resistin in direct ELISAs and Western blots. In these formats, this antibody shows less than 2% cross-reactivity with rmResistin, rmRELM alpha and rmRELM beta . | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Adipose tissue-specific secretory factor, ADSF, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN, RETN1, RSTNXCP1 | |
Goat | |
Affinity Purified | |
RUO | |
56729 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction