Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Resistin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB13591500
This item is not returnable.
View return policy
Description
Resistin Monoclonal specifically detects Resistin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Resistin | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Adipose tissue-specific secretory factor, ADSF, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN, RETN1, RSTNXCP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56729 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 184305 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RETN | |
| E. coli-derived recombinant human Resistin | |
| 500 μg | |
| Primary | |
| Detects human Resistin in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse RELM alpha . | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction