Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Respiratory Syncytial Virus Nucleoprotein VPN41 Antibody (4-18), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250535MFV610
Description
Human Respiratory Syncytial Virus Nucleoprotein VPN41 Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Nucleoprotein VPN41 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Human Respiratory Syncytial Virus Nucleoprotein VPN41 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Gradient-purified RSN-2 virus (subgroup B) treated with 0.1% SDS at 100C for 2 mins. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry | |
| 2024-04-18 00:00:00 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction