Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Rev-erb beta/NR1D2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH272900
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH272900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH272900 Supplier R&D Systems Supplier No. PPH272900
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rev-erb beta/NR1D2 Monoclonal specifically detects Rev-erb beta/NR1D2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA.

Specifications

Antigen Rev-erb beta/NR1D2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA
Classification Monoclonal
Clone H2729
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias BD73, EAR-1R, HZF2, NR1D2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, Reverb beta, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related
Gene Symbols NR1D2
Host Species Mouse
Immunogen Recombinant human Rev-erb beta/NR1D2 aa 4-101
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9975
Test Specificity This antibody specifically recognizes human Rev-Erb beta and cross-reacts with rat Rev-Erb beta . It does not recognize human Rev-Erb alpha . Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.