Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Rex-1/ZFP42 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3598
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3598 Supplier R&D Systems Supplier No. AF3598
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Rex-1/ZFP42 Polyclonal specifically detects Rex-1/ZFP42 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Rex-1/ZFP42
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. NP_777560
Gene Alias FLJ20086, KIAA1584, MGC21637, MGC61687, Rex1, SUHW4, Suppressor of hairy wing homolog 4, ZFP42, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Gene Symbols ZFP42
Host Species Sheep
Immunogen E. coli-derived recombinant human Rex-1/ZFP42 Ser2-Lys150 Accession # NP_777560
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132625
Test Specificity Detects human Rex-1/ZFP42 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human YYI is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.