Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RFX4 Full-length ORF (AAH30644.1, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959837
Click to view available options
Quantity:
10 ug
25 ug

Used for AP, Array, ELISA, WB-Re

This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X3, and X5. It has been shown to interact with itself as well as with regulatory factors X2 and X3, but it does not interact with regulatory factor X1. This protein may be a transcriptional repressor rather than a transcriptional activator. Three transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MNWAAFGGSEFFIPEGIQIDSRCPLSRNITEWYHYYGIAVKESSQYYDVMYSKKGAAWVSETGKKEVSKQTVAYSPRSKLGTLLPEFPNVKDLNLPASLPEEKVSTFIMMYRTHCQRILDTVIRANFDEVQSFLLHFWQGMPPHMLPVLGSSTVVNIVGVCDSILYKAISGVLMPTVLQALPDSLTQVIRKFAKQLDEWLKVALHDLPENLRNIKFELSRRFSQILRRQTSLNHLCQASRTVIHSADITFQMLEDWRNVDLNSITKQTLYTMEDSRDEHRKLITQLYQEFDHLLEEQSPIESYIEWLDTMVDRCVVKVAAKRQGSLKKVAQQFLLMWSCFGTRVIRDMTLHSAPSFGSFHLIHLMFDDYVLYLLESLHCQERANELMRAMKGEGSTAEVREEIILTEAAAPTPSPVPSFSPAKSATSVEVPPPSSPVSNPSPEYTGLSTTGAMQSYTWSLTYTVTTAAGSPAENSQQLPCMRNTHVPSSSVTHRIPVYPHREEHGYTGSYNYGSYGNQHPHPMQSQYPALPHDTAISGPLHYAPYHRSSAQYPFNSPTSRMEPCLMSSTPRLHPTPVTPRWPEVPSANTCYTSPSVHSARYGNSSDMYTPLTTRRNSEYEHMQHFPGFAYINGEASTGWAK

Specifications

Accession Number AAH30644.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5992
Molecular Weight (g/mol) 96.14kDa
Name RFX4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MNWAAFGGSEFFIPEGIQIDSRCPLSRNITEWYHYYGIAVKESSQYYDVMYSKKGAAWVSETGKKEVSKQTVAYSPRSKLGTLLPEFPNVKDLNLPASLPEEKVSTFIMMYRTHCQRILDTVIRANFDEVQSFLLHFWQGMPPHMLPVLGSSTVVNIVGVCDSILYKAISGVLMPTVLQALPDSLTQVIRKFAKQLDEWLKVALHDLPENLRNIKFELSRRFSQILRRQTSLNHLCQASRTVIHSADITFQMLEDWRNVDLNSITKQTLYTMEDSRDEHRKLITQLYQEFDHLLEEQSPIESYIEWLDTMVDRCVVKVAAKRQGSLKKVAQQFLLMWSCFGTRVIRDMTLHSAPSFGSFHLIHLMFDDYVLYLLESLHCQERANELMRAMKGEGSTAEVREEIILTEAAAPTPSPVPSFSPAKSATSVEVPPPSSPVSNPSPEYTGLSTTGAMQSYTWSLTYTVTTAAGSPAENSQQLPCMRNTHVPSSSVTHRIPVYPHREEHGYTGSYNYGSYGNQHPHPMQSQYPALPHDTAISGPLHYAPYHRSSAQYPFNSPTSRMEPCLMSSTPRLHPTPVTPRWPEVPSANTCYTSPSVHSARYGNSSDMYTPLTTRRNSEYEHMQHFPGFAYINGEASTGWAK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NYD-SP10
Common Name RFX4
Gene Symbol RFX4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.