Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RFX5 Partial ORF (NP_000440, 516 a.a. - 616 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959840
Click to view available options
Quantity:
10 ug
25 ug

Used for AP, Array, ELISA, WB-Re

A lack of MHC-II expression results in a severe immunodeficiency syndrome called MHC-II deficiency, or the bare lymphocyte syndrome (BLS; MIM 209920). At least 4 complementation groups have been identified in B-cell lines established from patients with BLS. The molecular defects in complementation groups B, C, and D all lead to a deficiency in RFX, a nuclear protein complex that binds to the X box of MHC-II promoters. The lack of RFX binding activity in complementation group C results from mutations in the RFX5 gene encoding the 75-kD subunit of RFX (Steimle et al., 1995). RFX5 is the fifth member of the growing family of DNA-binding proteins sharing a novel and highly characteristic DNA-binding domain called the RFX motif. Multiple alternatively spliced transcript variants have been found but the full-length natures of only two have been determined. [provided by RefSeq]

Sequence: ERPGPMGEAEKGAVLAQGQGDGTVSKGGRGPGSQHTKEAEDKIPLVPSKVSVIKGSRSQKEAFPLAKGEVDTAPQGNKDLKEHVLQSSLSQEHKDPKATPP

Specifications

Accession Number NP_000440
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5993
Molecular Weight (g/mol) 36.85kDa
Name RFX5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen ERPGPMGEAEKGAVLAQGQGDGTVSKGGRGPGSQHTKEAEDKIPLVPSKVSVIKGSRSQKEAFPLAKGEVDTAPQGNKDLKEHVLQSSLSQEHKDPKATPP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name RFX5
Gene Symbol RFX5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.