Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RFX6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7780
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7780 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7780 Supplier R&D Systems Supplier No. AF7780
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

RFX6 Polyclonal specifically detects RFX6 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen RFX6
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8HWS3
Gene Alias dJ955L16.1, DNA-binding protein RFX6, MGC33442, MTFS, Regulatory factor X 6, Regulatory factor X domain-containing protein 1, regulatory factor X, 6, RFXDC1, RFXDC1regulatory factor X domain containing 1
Gene Symbols RFX6
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human RFX6 Lys324-Thr511 Accession # Q8HWS3
Purification Method Immunogen affinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 222546
Test Specificity Detects human RFX6 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.