Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RGM-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | RGM-B |
---|---|
Clone | 398528 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
RGM-B Monoclonal specifically detects RGM-B in Human samples. It is validated for Western Blot.Specifications
RGM-B | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q6NW40 | |
285704 | |
Mouse myeloma cell line NS0-derived recombinant human RGM-B isoform 1 Gly46-Ser411 Accession # Q6NW40 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human RGM-B in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse RGM-B, recombinant human (rh) RGM-A, or rhRGM-C is observed. |
398528 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp434P228, Dragon, DRG11-responsive axonal guidance and outgrowth of neurite, RGM domain family, member B, RGMB | |
RGMB | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title