Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RGM-B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3630
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3630 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3630 Supplier R&D Systems Supplier No. AF3630
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

RGM-B Polyclonal specifically detects RGM-B in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen RGM-B
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q6NW40
Gene Alias DKFZp434P228, Dragon, DRG11-responsive axonal guidance and outgrowth of neurite, RGM domain family, member B, RGMB
Gene Symbols RGMB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human RGM-B isoform 1 Gly46-SerSer411 Accession # Q6NW40
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285704
Test Specificity Detects human RGM-B in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse RGM-B is observed and less than 1% cross-reactivity with recombinant human (rh) RGM-A and rhRGM-C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.