Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RGM-C/Hemojuvelin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB37201100
This item is not returnable.
View return policy
Description
RGM-C/Hemojuvelin Monoclonal specifically detects RGM-C/Hemojuvelin in Human samples. It is validated for ELISA.Specifications
| RGM-C/Hemojuvelin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DL-M, haemojuvelin, hemochromatosis type 2 (juvenile), hemojuvelin, HFE2, HFE2AMGC23953, HJV, HJVHemochromatosis type 2 protein, JH, repulsive guidance molecule c, RGMC, RGMCRGM domain family member C | |
| Mouse myeloma cell line, NS0-derived human RGM-C/Hemojuvelin, Gln36-Asp400, Accession No. Q6ZVN8 | |
| 100 μg | |
| 148738.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 751757 | |
| ELISA | |
| Q6ZVN8 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human RGM-C/Hemojuvelin in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction