Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rhesus Macaque IL-18/IL-1F4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2548
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2548 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2548 Supplier R&D Systems Supplier No. AF2548
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-18/IL-1F4 Polyclonal specifically detects IL-18/IL-1F4 in Human, Rhesus Macaque samples. It is validated for Western Blot, Simple Western, ELISA, Neutralization, Immunocytochemistry, Knockout Validated.

Specifications

Antigen IL-18/IL-1F4
Applications Western Blot, ELISA, Neutralization, Immunocytochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL, ELISA, Neutralization 0.4-1.2 μg/mL, Immunocytochemistry 5-15 μg/mL, Knockout Validated
Gene Accession No. AAK13416
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Gene Symbols IL18
Host Species Goat
Immunogen E. coli-derived recombinant rhesus macaque IL-18/IL-1F4 Tyr37-Asp193 Accession # AAK13416
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity Detects rhesus macaque IL-18/IL-1F4 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse IL-18, recombinant rat IL-18, and recombinant porcine IL-18 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Rhesus Macaque
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.