Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ribosomal Protein S6/RPS6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM5436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM5436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM5436 Supplier R&D Systems Supplier No. BAM5436
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ribosomal Protein S6

Ribosomal Protein S6/RPS6 Monoclonal specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Immunocytochemistry.

Order Info

The product is shipped at ambient temperature.

Specifications

Antigen Ribosomal Protein S6/RPS6
Applications Immunocytochemistry
Classification Monoclonal
Clone 522731
Concentration LYOPH
Conjugate Biotin
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. P62753
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6
Gene Symbols RPS6
Host Species Mouse
Immunogen E. coli-derived recombinant human Ribosomal Protein S6/RPS6 Met1-Lys249 Accession # P62753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6194
Test Specificity Conjugated Ribosomal Protein S6/RPS6 antibodies are ideal for immunocytochemistry colocalization studies in ribosomes. Detects human Ribosomal Protein S6/RPS6 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.