Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RIG-I Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4859
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4859 Supplier R&D Systems Supplier No. AF4859
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

RIG-I Polyclonal specifically detects RIG-I in Human samples. It is validated for Western Blot.

Specifications

Antigen RIG-I
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. O95786
Gene Alias DDX58, DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIGI, RIG-IDKFZp434J1111, RNA helicase RIG-I
Gene Symbols DDX58
Host Species Goat
Immunogen E. coli-derived recombinant human RIG-I Met724-Lys925 Accession # O95786
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23586
Test Specificity Detects human RIG-I in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.