Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human RIPK3/RIP3 ELISA Kit (Colorimetric)
SDP

Catalog No. NB171841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171841 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171841 Supplier Novus Biologicals™ Supplier No. NBP318218
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.16-10 ng/ml
Assay Sensitivity 0.08 ng/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Cell Lysates, Tissue Homogenates
For Use With (Application) ELISA
Gene ID (Entrez) 11035
Gene Symbol RIPK3
Quantity 1 Kit
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.