Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Rit1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4697SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4697SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4697SP Supplier R&D Systems Supplier No. MAB4697SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rit1 Monoclonal specifically detects Rit1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Rit1
Applications Western Blot
Classification Monoclonal
Clone 456201
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q92963
Gene Alias GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBB, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1, ROC1Ric (Drosophila)-like, expressed in many tissues
Gene Symbols RIT1
Host Species Mouse
Immunogen E. coli-derived recombinant human Rit1 Met1-Thr219 Accession # Q92963
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6016
Test Specificity Detects human Rit1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.