Learn More
Human RNF43 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | RNF43 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 923227R |
| Conjugate | Alexa Fluor 750 |
| Dilution | Flow Cytometry 0.25-1 μg/10^6 cells |
| Formulation | Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q68DV7 |
| Gene Alias | DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.