Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RNF43 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7964100
This item is not returnable.
View return policy
Description
RNF43 Monoclonal specifically detects RNF43 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
RNF43 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC | |
Human embryonic kidney cell line HEK293-derived recombinant human RNF43 Glu43-Val199 Accession # Q68DV7 | |
100 μg | |
54894 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
CyTOF, Flow Cytometry | |
923227R | |
Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
Q68DV7 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human RNF43 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction