Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RNPS1 Full-length ORF (NP_006702.1, 1 a.a. - 305 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098983
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. This protein contains many serine residues. Two splice variants have been found for this gene; both variants encode the same protein. [provided by RefSeq]

Sequence: MDLSGVKKKSLLGVKENNKKSSTRAPSPTKRKDRSDEKSKDRSKDKGATKESSEKDRGRDKTRKRRSASSGSSSTRSRSSSTSSSGSSTSTGSSSGSSSSSASSRSGSSSTSRSSSSSSSSGSPSPSRRRHDNRRRSRSKSKPPKRDEKERKRRSPSPKPTKVHIGRLTRNVTKDHIMEIFSTYGKIKMIDMPVERMHPHLSKGYAYVEFENPDEAEKALKHMDGGQIDGQEITATAVLAPWPRPPPRRFSPPRRMLPPPPMWRRSPPRMRRRSRSPRRRSPVRRRSRSPGRRRHRSRSSSNSSR

Specifications

Accession Number NP_006702.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10921
Molecular Weight (g/mol) 60.6kDa
Name RNPS1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MDLSGVKKKSLLGVKENNKKSSTRAPSPTKRKDRSDEKSKDRSKDKGATKESSEKDRGRDKTRKRRSASSGSSSTRSRSSSTSSSGSSTSTGSSSGSSSSSASSRSGSSSTSRSSSSSSSSGSPSPSRRRHDNRRRSRSKSKPPKRDEKERKRRSPSPKPTKVHIGRLTRNVTKDHIMEIFSTYGKIKMIDMPVERMHPHLSKGYAYVEFENPDEAEKALKHMDGGQIDGQEITATAVLAPWPRPPPRRFSPPRRMLPPPPMWRRSPPRMRRRSRSPRRRSPVRRRSRSPGRRRHRSRSSSNSSR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias E5.1/MGC117332
Common Name RNPS1
Gene Symbol RNPS1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.