Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ROBO1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA71181T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA71181T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA71181T100 Supplier R&D Systems Supplier No. FAB71181T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ROBO1 Monoclonal specifically detects ROBO1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ROBO1
Applications Flow Cytometry
Classification Monoclonal
Clone 770502
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9Y6N7
Gene Alias DUTT1, DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
Gene Symbols ROBO1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ROBO1 Met1-Ala858 Accession # Q9Y6N7
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6091
Test Specificity Detects human ROBO1 in ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.