Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROBO1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7118SP
This item is not returnable.
View return policy
Description
ROBO1 Polyclonal specifically detects ROBO1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
ROBO1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DUTT1, DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
Sheep | |
Affinity Purified | |
RUO | |
6091 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
NP_598334 | |
ROBO1 | |
Mouse myeloma cell line NS0-derived recombinant human ROBO1 Ser20-Pro861 (Val310-Ser312 del) Accession # NP_598334 | |
25 μg | |
Primary | |
Detects human ROBO1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant rat ROBO1 is observed, and less than 3% cross-reactivity with recombinant human (rh) ROBO2 and rhROBO3 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction