Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ROBO1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7118
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7118 Supplier R&D Systems Supplier No. AF7118
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ROBO1 Polyclonal specifically detects ROBO1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen ROBO1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. NP_598334
Gene Alias DUTT1, DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
Gene Symbols ROBO1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human ROBO1 Ser20-Pro861 (Val310-Ser312 del) Accession # NP_598334
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6091
Test Specificity Detects human ROBO1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant rat ROBO1 is observed, and less than 3% cross-reactivity with recombinant human (rh) ROBO2 and rhROBO3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.