Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROBO2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
$227.50 - $596.50
Specifications
Antigen | ROBO2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Polyclonal |
Description
ROBO2 Polyclonal specifically detects ROBO2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
ROBO2 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila), SAX3 | |
ROBO2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
Q9HCK4 | |
6092 | |
Mouse myeloma cell line NS0-derived recombinant human ROBO2 Ser22-Ala312, RPVA, Pro313-Pro859 Accession # Q9HCK4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ROBO2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO3 and rhROBO4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title