Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROR alpha/NR1F1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | ROR alpha/NR1F1 |
---|---|
Clone | 784652 |
Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Description
ROR alpha/NR1F1 Monoclonal specifically detects ROR alpha/NR1F1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
ROR alpha/NR1F1 | |
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp686M2414, MGC119326, NR1F1, NR1F1MGC119329, nuclear receptor ROR-alpha, Nuclear receptor RZR-alpha, Nuclear receptor subfamily 1 group F member 1, RAR-related orphan receptor A, RAR-related orphan receptor alpha, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, Retinoid-related orphan receptor-alpha, ROR1, ROR2, ROR3, RORA, RZRA, RZR-ALPHA, RZRAROR-alpha, transcription factor RZR-alpha | |
RORA | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
784652 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P35398 | |
6095 | |
E. coli-derived recombinant human ROR alpha/NR1F1 Arg47-His163 Accession # P35398 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ROR alpha/NR1F1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title