Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ROR-beta Antibody (Clone N7927), R&D Systems™
GREENER_CHOICE

Catalog No. PPN792700
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPN792700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPN792700 Supplier R&D Systems Supplier No. PPN792700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ROR beta Monoclonal specifically detects ROR beta in Human samples. It is validated for .

Specifications

Antigen ROR beta
Classification Monoclonal
Clone N7927
Conjugate Unconjugated
Gene Alias NR1F2, NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, RORB, ROR-BETA, RZRB, RZRBbA133M9.1, RZR-BETA
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6096
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.