Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROR gamma/RORC/NR1F3 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6109C
This item is not returnable.
View return policy
Description
ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
ROR gamma/RORC/NR1F3 | |
Monoclonal | |
PerCP | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
MGC129539, NR1F3, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORC, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6097 | |
Human | |
Purified |
Flow Cytometry | |
600214 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
P51449-2 | |
RORC | |
E. coli-derived recombinant human ROR gamma/RORC/NR1F3 Met1-Gln100 Accession # P51449-2 | |
100 Tests | |
Primary | |
Detects human and mouse ROR gamma/RORC/NR1F3 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction