Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ROR1 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB20001U10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB20001U10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB20001U10 Supplier R&D Systems Supplier No. FAB20001U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ROR1 Monoclonal specifically detects ROR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ROR1
Applications Flow Cytometry
Classification Monoclonal
Clone 1011919
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q01973
Gene Alias EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ROR1, Ala24-Glu403, Accession # Q01973
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4919
Test Specificity Detects human ROR1 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.