Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROR1 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB2000G
This item is not returnable.
View return policy
Description
ROR1 Polyclonal specifically detects ROR1 in Human samples. It is validated for Flow Cytometry.Specifications
ROR1 | |
Polyclonal | |
Flow Cytometry 5 uL/10^6 cells | |
EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
Goat | |
Affinity Purified | |
RUO | |
4919 | |
Human | |
IgG |
Flow Cytometry | |
Alexa Fluor 488 | |
Q01973 | |
ROR1 | |
Mouse myeloma cell line NS0-derived recombinant human ROR1 Ala24-Glu403 Accession # Q01973 | |
100 Tests | |
Primary | |
Detects human ROR1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human ROR2 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction