Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROR1 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB20001N100UG
This item is not returnable.
View return policy
Description
ROR1 Monoclonal specifically detects ROR1 in Human samples. It is validated for Flow Cytometry.Specifications
ROR1 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
Chinese hamster ovary cell line CHO-derived recombinant human ROR1, Ala24-Glu403, Accession # Q01973 | |
100 μg | |
4919 | |
Human | |
Supernatant |
Flow Cytometry | |
1011919 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q01973 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human ROR1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction