Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ROR2 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB20641A
This item is not returnable.
View return policy
Description
ROR2 Monoclonal specifically detects ROR2 in Human samples. It is validated for Flow Cytometry.Specifications
ROR2 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4920 | |
Human | |
Purified |
Flow Cytometry | |
231509 | |
Flow Cytometry 10 uL/10^6 cells | |
AAA60276 | |
ROR2 | |
Mouse myeloma cell line NS0-derived recombinant human ROR2 Val28-Gly403 Accession # AAA60276 | |
100 Tests | |
Primary | |
Detects human ROR2 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction