Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RPGRIP1 Partial ORF (NP_065099.2, 1187 a.a. - 1286 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101459
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a photoreceptor protein that interacts with retinitis pigmentosa GTPase regulator protein and is a key component of cone and rod photoreceptor cells. Mutations in this gene lead to autosomal recessive congenital blindness. [provided by RefSeq]

Sequence: QGRRRFLFDMLNGQDPDQGHLKFTVVSDPLDEEKKECEEVGYAYLQLWQILESGRDILEQELDIVSPEDLATPIGRLKVSLQAAAVLHAIYKEMTEDLFS

Specifications

Accession Number NP_065099.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57096
Molecular Weight (g/mol) 36.74kDa
Name RPGRIP1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen QGRRRFLFDMLNGQDPDQGHLKFTVVSDPLDEEKKECEEVGYAYLQLWQILESGRDILEQELDIVSPEDLATPIGRLKVSLQAAAVLHAIYKEMTEDLFS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CORD9/DKFZp686P0897/LCA6/RGI1/RGRIP/RPGRIP/RPGRIP1d
Common Name RPGRIP1
Gene Symbol RPGRIP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.