Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RPS6KA3 (NM_004586) Full-length Recombinant Protein with GST expressed in Sf9 cells

Catalog No. 89960744
Click to view available options
Quantity:
5 μg

Used for Func, SDS-PAGE

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in this gene have been associated with Coffin-Lowry syndrome (CLS). [provided by RefSeq]

Specifications

Accession Number NM_004586
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 6197
Molecular Weight (g/mol) 112kDa
Name RPS6KA3 (Human) Recombinant Protein
Quality Control Testing SDS-PAGE Result
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CLS/HU-3/ISPK-1/MAPKAPK1B/MRX19/RSK/RSK2/S6K-alpha3/p90-RSK2/pp90RSK2
Common Name RPS6KA3
Gene Symbol RPS6KA3
Biological Activity The specific activity of RSK2 was determined to be 157nmol/min/mg as per activity assay protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.