Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RRAGD Full-length ORF (NP_067067.1, 1 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960815
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.[supplied by OMIM]

Sequence: MSQVLGKPQPQDEDDAEEEEEEDELVGLADYGDGPDSSDADPDSGTEEGVLDFSDPFSTEVKPRILLMGLRRSGKSSIQKVVFHKMSPNETLFLESTNKICREDVSNSSFVNFQIWDFPGQIDFFDPTFDYEMIFRGTGALIFVIDSQDDYMEALARLHLTVTRAYKVNTDINFEVFIHKVDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLL

Specifications

Accession Number NP_067067.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 58528
Molecular Weight (g/mol) 72kDa
Name RRAGD (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MSQVLGKPQPQDEDDAEEEEEEDELVGLADYGDGPDSSDADPDSGTEEGVLDFSDPFSTEVKPRILLMGLRRSGKSSIQKVVFHKMSPNETLFLESTNKICREDVSNSSFVNFQIWDFPGQIDFFDPTFDYEMIFRGTGALIFVIDSQDDYMEALARLHLTVTRAYKVNTDINFEVFIHKVDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp761H171/FLJ44176/RAGD/bA11D8.2.1
Common Name RRAGD
Gene Symbol RRAGD
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.