Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RSK4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3916SP
This item is not returnable.
View return policy
Description
RSK4 Monoclonal specifically detects RSK4 in Human samples. It is validated for Western Blot.Specifications
RSK4 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q9UK32 | |
RPS6KA6 | |
E. coli-derived recombinant human RSK4 Met1-Leu745 Accession # Q9UK32 | |
25 μg | |
Primary | |
Detects human RSK4 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) RSK1, rhRSK2, or rhRSK3 is observed. | |
Human | |
Purified |
Western Blot | |
374521 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.11, EC 2.7.11.1, p90-RSK 6, p90RSK6, pp90RSK4, PP90RSK4,90 kDa ribosomal protein S6 kinase 6, ribosomal protein S6 kinase alpha-6, ribosomal protein S6 kinase, 90kDa, polypeptide 6, Ribosomal S6 kinase 4, RPS6KA6, RSK-4, RSK4ribosomal protein S6 kinase, 90kD, polypeptide 6, S6K-alpha 6, S6K-alpha-6 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27330 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction