Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RT28 (aa 1-92) Control Fragment Recombinant Protein

Catalog No. RP93813
Encompass_Preferred
Click to view available options
Quantity:
100 μL

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55542 (PA5-55542. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that has been called mitochondrial ribosomal protein S35 in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y2Q9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 28957
Name Human RT28 (aa 1-92) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500012D08Rik; 28 S ribosomal protein S28, mitochondrial; 28 S ribosomal protein S35, mitochondrial; AW061224; HSPC007; LRRGT00031; mitochondrial 28 S ribosomal protein S35; mitochondrial ribosomal protein S28; mitochondrial ribosomal protein S28-like; Mitochondrial small ribosomal subunit protein bS1m; Mrps28; MRP-S28; Mrps28l; MRPS35; MRP-S35; S28mt; S35mt
Common Name RT28
Gene Symbol MRPS28
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAALCRTRAVAAESHFLRVFLFFRPFRGVGTESGSESGSSNAKEPKTRAGGFASALERHSELLQKVEPLQKGSPKNVESFASMLRHSPLTQM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.