Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RTTN Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $491.00
Specifications
Antigen | rotatin |
---|---|
Clone | 2311A |
Dilution | Western Blot 0.5 ug/mL, ELISA, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9966SP
![]() |
R&D Systems
MAB9966SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9966100
![]() |
R&D Systems
MAB9966100 |
100 μg |
Each for $491.00
|
|
|||||
Description
rotatin Monoclonal specifically detects rotatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.Specifications
rotatin | |
Western Blot 0.5 ug/mL, ELISA, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DKFZp434G145, FLJ26356, FLJ39085, MSSP, PMGYS, rotatin | |
Synthetic peptide containing human RTTN Accession # Q86VV8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human RTTN in direct ELISAs. |
2311A | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Human | |
Q86VV8 | |
25914 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title