Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RUNX1/CBFA2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2399
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2399 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2399 Supplier R&D Systems Supplier No. BAF2399
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

RUNX1/CBFA2 Polyclonal specifically detects RUNX1/CBFA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen RUNX1/CBFA2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q01196
Gene Alias Acute myeloid leukemia 1 protein, AML1, AML1acute myeloid leukemia 1, AML1-EVI-1, CBFA2, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, Core-binding factor subunit alpha-2, EVI-1, PEBP2A2, runt domain, alpha subunit 2, runt-related transcription factor 1, SL3/AKV core-binding factor alpha B subunit
Gene Symbols RUNX1
Host Species Goat
Immunogen E. coli-derived recombinant human RUNX1/CBFA2 Gly190-Tyr453 Accession # Q01196
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 861
Test Specificity Detects human RUNX1/CBFA2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.