Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RXR alpha/NR2B1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPK850800
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPK850800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPK850800 Supplier R&D Systems Supplier No. PPK850800
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RXR alpha/NR2B1 Monoclonal specifically detects RXR alpha/NR2B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Supershift Assay.

Specifications

Antigen RXR alpha/NR2B1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Shift
Classification Monoclonal
Clone K8508
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Supershift Assay
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha, RXRA
Gene Symbols RXRA
Host Species Mouse
Immunogen Recombinant human RXRα (amino acids 2 - 133) and mouse myeloma cells (NS-1)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6256
Target Species Human
Content And Storage Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.