Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ryk Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4907
This item is not returnable.
View return policy
Description
Ryk Polyclonal specifically detects Ryk in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Ryk | |
Polyclonal | |
Unconjugated | |
NP_001005861 | |
RYK | |
Mouse myeloma cell line NS0-derived recombinant human Ryk Ala47-Thr224 Accession # NP_001005861 | |
100 μg | |
Primary | |
Detects human Ryk in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse Ryk is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
D3S3195, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, JTK5, JTK5A, JTK5A protein tyrosine kinase, RYK receptor-like tyrosine kinase, RYK1, tyrosine-protein kinase RYK | |
Sheep | |
Affinity Purified | |
RUO | |
6259 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction