Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human S100A11 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4874SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4874SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4874SP Supplier R&D Systems Supplier No. AF4874SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

S100A11 Polyclonal specifically detects S100A11 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen S100A11
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P31949
Gene Alias calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin)
Gene Symbols S100A11
Host Species Goat
Immunogen E. coli-derived recombinant human S100A11 Ala2-Thr105 Accession # P31949
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6282
Test Specificity Detects human S100A11 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) S100A1, A2, A7, A8, A10, A13, A16, rhS100B, rhS100P, recombinant mouse (rm) S100A6, and rmS100A10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.