Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S100A11 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4874SP
This item is not returnable.
View return policy
Description
S100A11 Monoclonal specifically detects S100A11 in Human samples. It is validated for Western Blot.Specifications
S100A11 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P31949 | |
S100A11 | |
E. coli-derived recombinant human S100A11 Ala2-Thr105 Accession # P31949 | |
25 μg | |
Primary | |
Detects human S100A11 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) S100A1, A2, A4, A6, A7, A8, A9, A10, A13, rhS100B, rhS100P, recombinant mouse S100A1, A4, A6, A8, A9, A10, A11, A13, or A16 is observed. | |
Human | |
Purified |
Western Blot | |
672816 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6282 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction