Learn More
Human S100A16 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | S100A16 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gene Accession No. | Q96FQ6 |
| Gene Alias | AAG13, Aging-associated gene 13 protein, aging-associated protein 13, DT1P1A7, MGC17528, protein S100-A16, Protein S100-F, S100 calcium binding protein A16, S100F, S100FS100 calcium-binding protein A16 |
| Gene Symbols | S100A16 |
| Host Species | Sheep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.