Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S100A2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4870
This item is not returnable.
View return policy
Description
S100A2 Polyclonal specifically detects S100A2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
S100A2 | |
Polyclonal | |
Unconjugated | |
P29034 | |
S100A2 | |
E. coli-derived recombinant human S100A2 Met2-Pro98 Accession # P29034 | |
100 μg | |
Primary | |
Detects human S100A2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) S100A1, -A6, -A7, -A8, -A10, -A11, -A13, -A16, rhS100B and rhS100P is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
CAN19, CAN19protein S100-A2, MGC111539, S100 calcium binding protein A2, S100 calcium-binding protein A2Protein S-100L, S100L | |
Goat | |
Affinity Purified | |
RUO | |
6273 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction