Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human S100A8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4570
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4570 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4570 Supplier R&D Systems Supplier No. AF4570
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

S100A8 Polyclonal specifically detects S100A8 in Human samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen S100A8
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. P05109
Gene Alias 60B8AG, CAGACP-10, calgranulin A, Calgranulin-A, Calprotectin L1L subunit, CFAG, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Gene Symbols S100A8
Host Species Sheep
Immunogen E. coli-derived recombinant human S100A8 Met1-Glu93 Accession # P05109
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6279
Test Specificity Detects human S100A8 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant mouse S100A8, recombinant human (rh) S100A1, and rhS100P is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.