Learn More
Human S100A8/S100A9 Heterodimer Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | S100A8/S100A9 Heterodimer |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 900028 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P05109 (S100A8) and P06702 (S100A9) |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9), Accession No. P05109 (S100A8) and P06702 (S100A9) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.