Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S100A8/S100A9 Heterodimer Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC45702S100UG
This item is not returnable.
View return policy
Description
S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
S100A8/S100A9 Heterodimer | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Human | |
Supernatant |
Flow Cytometry | |
900027 | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
P05109 (S100A8) and P06702 (S100A9) | |
E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9), Accession No. P05109 (S100A8) and P06702 (S100A9) | |
100 μg | |
Detects human S100A8/S100A9 heterodimer in direct ELISAs. In direct ELISAs, no detection of recombinant human S100A8 or S100A9 monomers was observered. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction