Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human S100A8/S100A9 Heterodimer Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB45701100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB45701100 100 μg
MAB45701SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB45701100 Supplier R&D Systems Supplier No. MAB45701100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Dual RNAscope ISH-IHC.

Specifications

Antigen S100A8/S100A9 Heterodimer
Applications Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH)
Classification Monoclonal
Clone 1099F
Conjugate Unconjugated
Dilution Immunohistochemistry 10-25 ug/mL, Immunocytochemistry 3-25 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. P05109 (S100A8) and P06702 (S100A9)
Host Species Rabbit
Immunogen E. coli-derived recombinant human S100A8/S100A9 heterodimer Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9) Accession # P05109 (S100A8) and P06702 (S100A9)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human S100A8/S100A9 in direct ELISAs. In ELISAs, it detects recombinant human S100A8/S100A9 heterodimer but it does not detect recombinant human S100A8 or S100A9 monomers.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.