Learn More
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Binding Target | S100A8/S100A9 Heterodimer |
| For Use With (Application) | ELISA |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Immunology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
