Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S100B Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1820
This item is not returnable.
View return policy
Description
S100B Polyclonal specifically detects S100B in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
S100B | |
Polyclonal | |
Unconjugated | |
P04271 | |
S100B | |
E. coli-derived recombinant human S100B Met1-Glu92 Accession # P04271 | |
100 μg | |
Primary | |
Detects human S100B in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) S100A1, rhS100A4, and rhS100A6 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
Goat | |
Affinity Purified | |
RUO | |
6285 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction